Product Details

SNP ID
rs717768
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:17320785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACCAAATTTTGGGGCTCATCTCA[A/G]AATACTCTTTAGCCACCTCCTTTGG
Phenotype
MIM: 610139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA6 PubMed Links

Gene Details

Gene
ST8SIA6
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004470.1 1282 UTR 3 NP_001004470.1
XM_017016174.1 1282 UTR 3 XP_016871663.1
XM_017016175.1 1282 Intron XP_016871664.1

View Full Product Details