Product Details

SNP ID
rs461126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87148659 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGGCGGCGCGGCAGCCAGCGAG[A/C]GGGGAGAAACAGATTCACACCCAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9orf170 PubMed Links

Gene Details

Gene
C9orf170
Gene Name
chromosome 9 open reading frame 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001709.2 16 UTR 5 NP_001001709.1

View Full Product Details