Product Details

SNP ID
rs462920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:87150143 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTTTGTCGGTCCCCCATAGTCCCC[A/G]ATCCCTCTAGCACTTTGGTTTCATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf170 PubMed Links

Gene Details

Gene
C9orf170
Gene Name
chromosome 9 open reading frame 170
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001709.2 Intron NP_001001709.1

View Full Product Details