Product Details

SNP ID
rs561630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135705600 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGTGCTAGGTGGCTGGGTGCCAAG[A/G]CCCGGTTAAATGGGCATCTTTCAAA
Phenotype
MIM: 608167 MIM: 610224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNT1 PubMed Links

Gene Details

Gene
KCNT1
Gene Name
potassium sodium-activated channel subfamily T member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272003.1 Intron NP_001258932.1
NM_020822.2 Intron NP_065873.2
XM_011518877.2 Intron XP_011517179.1
XM_011518878.2 Intron XP_011517180.1
XM_011518879.2 Intron XP_011517181.1
XM_011518880.1 Intron XP_011517182.1
XM_011518881.2 Intron XP_011517183.1
XM_017014931.1 Intron XP_016870420.1
XM_017014932.1 Intron XP_016870421.1
XM_017014933.1 Intron XP_016870422.1
Gene
SOHLH1
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
There are no transcripts associated with this gene.

View Full Product Details