Product Details
- SNP ID
-
rs240229
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:80005475 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGTTATTCCCATTGCCAAACGAATT[A/C]TTTATGTCTATGTCACTACCAGAAT
- Phenotype
-
MIM: 604092
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
TTK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9341801] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TTK
- Gene Name
- TTK protein kinase
View Full Product Details