Product Details
- SNP ID
-
rs403350
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:8047922 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GGGCACAAACATGGATTTTTTTTTT[C/T]ACCAGGTGGTCTCTAGCATGTCCAA
- Phenotype
-
MIM: 131320
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GATA3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76572342] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GATA3
- Gene Name
- GATA binding protein 3
- Gene
- GATA3-AS1
- Gene Name
- GATA3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details