Product Details

SNP ID
rs663632
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119153756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACAGCTTTGATCCCCTCTCCAC[C/G]CCCATCCATGAGATGTGTGGATTTT
Phenotype
MIM: 607784
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
ABCG4 PubMed Links

Gene Details

Gene
ABCG4
Gene Name
ATP binding cassette subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142505.1 544 Intron NP_001135977.1
NM_022169.4 544 Intron NP_071452.2
XM_011542952.2 544 UTR 5 XP_011541254.1
XM_017018121.1 544 Intron XP_016873610.1

View Full Product Details