Product Details

SNP ID
rs540018
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135693075 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTGCAGAGATGACCAGCAGTGGA[A/G]GTGGAAGCCAGCCCTTCCCTTGGAG
Phenotype
MIM: 608167 MIM: 610224
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNT1 PubMed Links

Gene Details

Gene
KCNT1
Gene Name
potassium sodium-activated channel subfamily T member 1
There are no transcripts associated with this gene.

Gene
SOHLH1
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012415.2 2889 Intron NP_001012415.2
NM_001101677.1 2889 Intron NP_001095147.1
XM_005263403.3 2889 UTR 3 XP_005263460.1
XM_006717109.3 2889 UTR 3 XP_006717172.1
XM_011518698.2 2889 UTR 3 XP_011517000.1

View Full Product Details