Product Details
- SNP ID
-
rs25664
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120212461 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGCCATGGCGGTGATGCAGCGGCC[A/G]GTGATGGGCTTCTGGCAGCCAGAAC
- Phenotype
-
MIM: 602505
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PXN
PubMed Links
Gene Details
- Gene
- PXN
- Gene Name
- paxillin
- Gene
- PXN-AS1
- Gene Name
- PXN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details