Product Details

SNP ID
rs41416
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:72177879 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGATGCACAATTATACTCTTGCCA[C/G]TAGCCATCTTTCTGCTACTAGCCAG
Phenotype
MIM: 607880
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EXOC6B PubMed Links

Gene Details

Gene
EXOC6B
Gene Name
exocyst complex component 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321729.1 3328 UTR 3 NP_001308658.1
NM_001321730.1 3328 UTR 3 NP_001308659.1
NM_001321731.1 3328 UTR 3 NP_001308660.1
NM_001321733.1 3328 UTR 3 NP_001308662.1
NM_001321734.1 3328 UTR 3 NP_001308663.1
NM_015189.2 3328 UTR 3 NP_056004.1
XM_005264224.1 3328 UTR 3 XP_005264281.1
XM_011532711.2 3328 Intron XP_011531013.1
XM_011532712.2 3328 Intron XP_011531014.1
XM_017003641.1 3328 UTR 3 XP_016859130.1
XM_017003642.1 3328 UTR 3 XP_016859131.1

View Full Product Details