Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_002031.2 | Intron | NP_002022.1 | |||
XM_005266880.4 | Intron | XP_005266937.1 | |||
XM_005266881.2 | Intron | XP_005266938.1 | |||
XM_005266882.4 | Intron | XP_005266939.1 | |||
XM_011535653.2 | Intron | XP_011533955.1 | |||
XM_011535654.2 | Intron | XP_011533956.1 | |||
XM_011535655.2 | Intron | XP_011533957.1 | |||
XM_011535656.2 | Intron | XP_011533958.1 | |||
XM_017010645.1 | Intron | XP_016866134.1 |