Product Details

SNP ID
rs11773214
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:33095614 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGAATGCCTACTTTACGTGACTG[A/T]AACCAGTGTACAAGCCACTTTGTAT
Phenotype
MIM: 607331
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RP9 PubMed Links

Gene Details

Gene
RP9
Gene Name
retinitis pigmentosa 9 (autosomal dominant)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_203288.1 Intron NP_976033.1
XM_011515468.2 Intron XP_011513770.1

View Full Product Details