Product Details

SNP ID
rs1566496
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:168897916 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTAACAGCTGCAGAGCTGAGGAGG[A/C]GGCCCTTGCAAGGCAGGGATTGTTA
Phenotype
MIM: 608092
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CBR4 PubMed Links

Gene Details

Gene
CBR4
Gene Name
carbonyl reductase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032783.4 Intron NP_116172.2
XM_005263315.2 Intron XP_005263372.1
XM_005263316.3 Intron XP_005263373.1
XM_006714391.1 Intron XP_006714454.1
XM_006714392.3 Intron XP_006714455.1
XM_011532385.1 Intron XP_011530687.1
XM_011532386.2 Intron XP_011530688.1
XM_017008782.1 Intron XP_016864271.1
XM_017008783.1 Intron XP_016864272.1
Gene
PALLD
Gene Name
palladin, cytoskeletal associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166108.1 Intron NP_001159580.1
NM_001166109.1 Intron NP_001159581.1
NM_001166110.1 Intron NP_001159582.1
NM_016081.3 Intron NP_057165.3
XM_005262861.4 Intron XP_005262918.1
XM_005262866.2 Intron XP_005262923.1
XM_011531768.2 Intron XP_011530070.1
XM_011531769.2 Intron XP_011530071.1
XM_011531770.2 Intron XP_011530072.1
XM_011531771.2 Intron XP_011530073.1
XM_011531772.2 Intron XP_011530074.1
XM_011531773.1 Intron XP_011530075.1
XM_011531774.1 Intron XP_011530076.1
XM_011531775.1 Intron XP_011530077.1
XM_017007910.1 Intron XP_016863399.1
XM_017007911.1 Intron XP_016863400.1

View Full Product Details