Product Details

SNP ID
rs9634174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:119592576 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACTGCATGATCAAATTATTTTCA[C/T]TTCTTTGTGGTCCTGCAGCTCAGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105370027 PubMed Links

Gene Details

Gene
LOC105370027
Gene Name
uncharacterized LOC105370027
There are no transcripts associated with this gene.

Gene
TMEM233
Gene Name
transmembrane protein 233
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136534.1 454 Intron NP_001130006.1
XM_005253880.3 454 UTR 5 XP_005253937.1
XM_005253881.3 454 UTR 5 XP_005253938.1
XM_011538328.2 454 UTR 5 XP_011536630.1
XM_011538329.2 454 UTR 5 XP_011536631.1
XM_011538330.2 454 UTR 5 XP_011536632.1
XM_011538331.2 454 UTR 5 XP_011536633.1

View Full Product Details