Product Details

SNP ID
rs2153049
Assay Type
Functionally tested
NCBI dbSNP Submissions
48
Location
Chr.1:230911646 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACACAAGAATAATCTACATCAAGAA[A/G]AGGATTGTTTTCTTCCTCCCCCATA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TTC13 PubMed Links

Gene Details

Gene
TTC13
Gene Name
tetratricopeptide repeat domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122835.2 Intron NP_001116307.2
NM_024525.4 Intron NP_078801.3
XM_005273260.1 Intron XP_005273317.1
XM_005273261.2 Intron XP_005273318.1
XM_005273262.1 Intron XP_005273319.1
XM_005273264.2 Intron XP_005273321.1
XM_006711814.2 Intron XP_006711877.1
XM_006711815.2 Intron XP_006711878.1
XM_011544275.2 Intron XP_011542577.1
XM_011544276.2 Intron XP_011542578.1
XM_017002321.1 Intron XP_016857810.1
XM_017002322.1 Intron XP_016857811.1
XM_017002323.1 Intron XP_016857812.1
XM_017002324.1 Intron XP_016857813.1
XM_017002325.1 Intron XP_016857814.1
XM_017002326.1 Intron XP_016857815.1

View Full Product Details