Product Details

SNP ID
rs3922634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89510358 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACCTATTGCTCAGACTATTACTT[C/T]ATATTTCAGGGCAATGTAGATATTA
Phenotype
MIM: 602783
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPG7 PubMed Links

Gene Details

Gene
SPG7
Gene Name
SPG7, paraplegin matrix AAA peptidase subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003119.3 Intron NP_003110.1
NM_199367.2 Intron NP_955399.1
XM_005256321.4 Intron XP_005256378.1
XM_006721264.3 Intron XP_006721327.1
XM_017023597.1 Intron XP_016879086.1
XM_017023598.1 Intron XP_016879087.1
XM_017023599.1 Intron XP_016879088.1

View Full Product Details