Product Details

SNP ID
rs12747487
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:159407700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTATATTTCTCTATGAAATCAG[C/T]GATAAAATTCACAACTGATCTTAAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR10J1 PubMed Links

Gene Details

Gene
OR10J1
Gene Name
olfactory receptor family 10 subfamily J member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012351.2 Intron NP_036483.2
XM_006711271.2 Intron XP_006711334.1
XM_017001015.1 Intron XP_016856504.1

View Full Product Details