Product Details

SNP ID
rs2742470
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8982455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATTCTGGCCTTGCTCTTTTTTT[C/T]CCCCCATCTTTCTCTACTCCTGGTT
Phenotype
MIM: 613125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NRIP3 PubMed Links

Gene Details

Gene
NRIP3
Gene Name
nuclear receptor interacting protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020645.2 1930 UTR 3 NP_065696.1
Gene
TMEM9B-AS1
Gene Name
TMEM9B antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details