Product Details

SNP ID
rs4144165
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:99399384 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ACAAAATCAGACAATTTTAATGTCA[G/T]GAAATTGACACAAACCTGCATAATA
Phenotype
MIM: 605196 MIM: 616653
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COQ3 PubMed Links

Gene Details

Gene
COQ3
Gene Name
coenzyme Q3, methyltransferase
There are no transcripts associated with this gene.

Gene
PNISR
Gene Name
PNN interacting serine and arginine rich protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322405.1 3736 UTR 3 NP_001309334.1
NM_001322406.1 3736 UTR 3 NP_001309335.1
NM_001322408.1 3736 UTR 3 NP_001309337.1
NM_001322410.1 3736 UTR 3 NP_001309339.1
NM_001322412.1 3736 UTR 3 NP_001309341.1
NM_001322413.1 3736 UTR 3 NP_001309342.1
NM_001322414.1 3736 UTR 3 NP_001309343.1
NM_001322415.1 3736 UTR 3 NP_001309344.1
NM_001322416.1 3736 UTR 3 NP_001309345.1
NM_001322417.1 3736 Intron NP_001309346.1
NM_001322418.1 3736 Intron NP_001309347.1
NM_001322419.1 3736 Intron NP_001309348.1
NM_015491.2 3736 UTR 3 NP_056306.1
NM_032870.3 3736 UTR 3 NP_116259.2
XM_005266912.3 3736 UTR 3 XP_005266969.2
XM_017010710.1 3736 UTR 3 XP_016866199.1
XM_017010711.1 3736 UTR 3 XP_016866200.1
XM_017010712.1 3736 UTR 3 XP_016866201.1
XM_017010713.1 3736 UTR 3 XP_016866202.1

View Full Product Details