Product Details

SNP ID
rs35890195
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:156726811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTCTCCTCCCTCCCTCCACAAAC[A/G]TTGGTGTCTTCCCATAGTGATATCC
Phenotype
MIM: 611930
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ISG20L2 PubMed Links

Gene Details

Gene
ISG20L2
Gene Name
interferon stimulated exonuclease gene 20 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303095.1 Intron NP_001290024.1
XM_005245516.4 Intron XP_005245573.1
XM_005245518.4 Intron XP_005245575.1
XM_006711560.3 Intron XP_006711623.1
XM_006711561.3 Intron XP_006711624.1
Gene
RRNAD1
Gene Name
ribosomal RNA adenine dimethylase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details