Product Details
- SNP ID
-
rs28685844
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137307152 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CACCGGGACCCCTGCCAGCTGCAGC[C/T]GGGTGCCGTCGGCCAGCATGTCCGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EXD3
PubMed Links
Gene Details
- Gene
- EXD3
- Gene Name
- exonuclease 3'-5' domain containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001286823.1 |
3234 |
Intron |
|
|
NP_001273752.1 |
NM_017820.4 |
3234 |
Missense Mutation |
CAG,CGG |
Q810R |
NP_060290.3 |
XM_011518810.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q965R |
XP_011517112.1 |
XM_011518814.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q521R |
XP_011517116.1 |
XM_011518815.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q521R |
XP_011517117.1 |
XM_017014861.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q1014R |
XP_016870350.1 |
XM_017014862.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q928R |
XP_016870351.1 |
XM_017014863.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q914R |
XP_016870352.1 |
XM_017014864.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q904R |
XP_016870353.1 |
XM_017014865.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q904R |
XP_016870354.1 |
XM_017014866.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q793R |
XP_016870355.1 |
XM_017014867.1 |
3234 |
Missense Mutation |
CAG,CGG |
Q521R |
XP_016870356.1 |
- Gene
- NRARP
- Gene Name
- NOTCH-regulated ankyrin repeat protein
There are no transcripts associated with this gene.
View Full Product Details