Product Details

SNP ID
rs10027691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:155173925 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAGTCTAAATTCATGCATGAAAAT[A/G]TTTCTACCTCCAGTGCTTATGTACT
Phenotype
MIM: 162642
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NPY2R PubMed Links

Gene Details

Gene
NPY2R
Gene Name
neuropeptide Y receptor Y2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000910.3 Intron NP_000901.1
XM_005263033.4 Intron XP_005263090.1
XM_005263034.4 Intron XP_005263091.1

View Full Product Details