Product Details

SNP ID
rs2095858
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:35616586 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CGTTCGGTGTAAGTGGGAAGTAGGG[A/T]CAGCCTTACTCACAGCGCACTCCCA
Phenotype
MIM: 107272 MIM: 601782
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD72 PubMed Links

Gene Details

Gene
CD72
Gene Name
CD72 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001782.2 Intron NP_001773.1
XM_006716893.2 Intron XP_006716956.1
XM_006716894.3 Intron XP_006716957.1
XM_011518074.2 Intron XP_011516376.1
XM_017015351.1 Intron XP_016870840.1
Gene
MIR4667
Gene Name
microRNA 4667
There are no transcripts associated with this gene.

Gene
TESK1
Gene Name
testis-specific kinase 1
There are no transcripts associated with this gene.

View Full Product Details