Product Details

SNP ID
rs4782466
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:89096948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCACAGGCAGCGCTCTGGATTCC[A/G]CGCAGGAGAACAGCAGTTTCTTCTC
Phenotype
MIM: 614245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACSF3 PubMed Links

Gene Details

Gene
ACSF3
Gene Name
acyl-CoA synthetase family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127214.3 Intron NP_001120686.1
NM_001243279.2 Intron NP_001230208.1
NM_001284316.1 Intron NP_001271245.1
NM_174917.4 Intron NP_777577.2
XM_005256293.2 Intron XP_005256350.1
XM_011522942.1 Intron XP_011521244.1
XM_011522943.1 Intron XP_011521245.1
XM_017023018.1 Intron XP_016878507.1
XM_017023019.1 Intron XP_016878508.1
XM_017023020.1 Intron XP_016878509.1
XM_017023021.1 Intron XP_016878510.1
XM_017023022.1 Intron XP_016878511.1

View Full Product Details