Product Details

SNP ID
rs17201864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:88877743 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAACCCTCCAGAAGCCCACAAAAC[C/T]GCAAATGGCCCAGGGGAGCAAGCAT
Phenotype
MIM: 155760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACAN PubMed Links

Gene Details

Gene
ACAN
Gene Name
aggrecan
There are no transcripts associated with this gene.

Gene
HAPLN3
Gene Name
hyaluronan and proteoglycan link protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307952.1 1529 UTR 3 NP_001294881.1
NM_178232.3 1529 UTR 3 NP_839946.1
XM_011521261.1 1529 UTR 3 XP_011519563.1
XM_017021934.1 1529 UTR 3 XP_016877423.1
XM_017021935.1 1529 UTR 3 XP_016877424.1
XM_017021936.1 1529 UTR 3 XP_016877425.1

View Full Product Details