Product Details

SNP ID
rs12035608
Assay Type
Validated
NCBI dbSNP Submissions
66
Location
Chr.1:205572387 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGCCTCACCTTGGCCTCTCCCCTG[A/C]CCATGCTTTCTTAACAGAGGGTTTG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
MFSD4A PubMed Links

Gene Details

Gene
MFSD4A
Gene Name
major facilitator superfamily domain containing 4A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181644.4 Intron NP_857595.3
XM_011509229.2 Intron XP_011507531.1

View Full Product Details