Product Details
- SNP ID
-
rs10933528
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241012034 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGGGCCCGAGGCCTCTGCACACC[A/G]CACACCTGCAAGCTAGGCTCCCACT
- Phenotype
-
MIM: 616634
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SNED1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79073038] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNED1
- Gene Name
- sushi, nidogen and EGF like domains 1
View Full Product Details