Product Details

SNP ID
rs4098360
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:7681435 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CCTGGCAACGTGGGGCTAACATGGG[C/T]ATGATAGACTGTATAAAGGCAAGAG
Phenotype
MIM: 608342
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYB5R2 PubMed Links

Gene Details

Gene
CYB5R2
Gene Name
cytochrome b5 reductase 2
There are no transcripts associated with this gene.

Gene
OVCH2
Gene Name
ovochymase 2 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198185.4 Intron NP_937828.3
XM_017017659.1 Intron XP_016873148.1

View Full Product Details