Product Details

SNP ID
rs35013934
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:26697880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGAGGGGGCGCTGGGGGCTCA[G/T]GTTAGGAGTGTAGGCAGTGGTAGGA
Phenotype
MIM: 603024
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARID1A PubMed Links

Gene Details

Gene
ARID1A
Gene Name
AT-rich interaction domain 1A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006015.4 Intron NP_006006.3
NM_139135.2 Intron NP_624361.1
Gene
LOC101928728
Gene Name
uncharacterized LOC101928728
There are no transcripts associated with this gene.

View Full Product Details