Product Details

SNP ID
rs12544554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23247755 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTCAGAGAGATCAAGTGACTTAC[C/T]CAGAGTTACACAGCTAGTTAAAGGT
Phenotype
MIM: 611130
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHMP7 PubMed Links

Gene Details

Gene
CHMP7
Gene Name
charged multivesicular body protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317899.1 Intron NP_001304828.1
NM_152272.4 Intron NP_689485.1
XM_006716415.2 Intron XP_006716478.1
XM_017013961.1 Intron XP_016869450.1
XM_017013962.1 Intron XP_016869451.1
XM_017013963.1 Intron XP_016869452.1
XM_017013964.1 Intron XP_016869453.1
XM_017013965.1 Intron XP_016869454.1

View Full Product Details