Product Details

SNP ID
rs72552300
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:38352763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGAAATGGTCACAACTTCATGGTT[C/T]GAAATTTTCGGTAAGTGATGGTCAG
Phenotype
MIM: 300461
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OTC PubMed Links

Gene Details

Gene
OTC
Gene Name
ornithine carbamoyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000531.5 236 Nonsense Mutation CGA,TGA R23* NP_000522.3
XM_017029556.1 236 Nonsense Mutation CGA,TGA R23* XP_016885045.1

View Full Product Details