Product Details

SNP ID
rs316625
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:84303178 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCACTTTCTAAGAATTAAACA[C/T]AAGCTTTTCTCCCTTATTGATTAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SAMD13 PubMed Links

Gene Details

Gene
SAMD13
Gene Name
sterile alpha motif domain containing 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010971.2 Intron NP_001010971.1
NM_001134663.1 Intron NP_001128135.1
NM_001134664.1 Intron NP_001128136.1
XM_017000377.1 Intron XP_016855866.1

View Full Product Details