Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sesn-1
Entrez Gene ID
171593
Gene Name
Sestrin homolog
Gene Aliases
CELE_Y74C9A.5
Chromosome Location
Chr.I: 27591 - 32548 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058263.5 NP_490664.2 1 - 2 134 144

View Full Product Details