Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
col-45
Entrez Gene ID
171605
Gene Name
protein-coding
Gene Aliases
CELE_F53G12.7
Chromosome Location
Chr.I: 115687 - 117498 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306335.3 NP_001293264.1 1 - 2 206 86
NM_058278.4 NP_490679.1 2 - 3 465 86

View Full Product Details