Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
syg-2
Entrez Gene ID
181561
Gene Name
protein-coding
Gene Aliases
CELE_C26G2.1
Chromosome Location
Chr.X: 14654968 - 14662865 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322663.4 NP_001309674.1 22 - 23 3551 78
NM_001322664.3 NP_001309467.1 21 - 22 3512 78
NM_001322665.4 NP_001309678.1 17 - 18 2623 78
NM_001322667.4 NP_001309677.1 11 - 12 2086 78
NM_001322668.4 NP_001309676.1 11 - 12 1999 78
NM_001322669.4 NP_001309675.1 2 - 3 124 78
NM_001383675.1 21 - 22 3505 78

View Full Product Details