Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
slt-1
Entrez Gene ID
181562
Gene Name
Slit homolog 1 protein
Gene Aliases
CELE_F40E10.4
Chromosome Location
Chr.X: 14674328 - 14683332 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078036.7 NP_510437.2 29 - 30 4021 82

View Full Product Details