Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atrn-1
Entrez Gene ID
181566
Gene Name
Putative protein tag-53
Gene Aliases
CELE_F33C8.1
Chromosome Location
Chr.X: 14711579 - 14718787 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078042.6 NP_510443.5 1 - 2 130 84

View Full Product Details