Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tsp-8
Entrez Gene ID
181567
Gene Name
Tetraspanin
Gene Aliases
CELE_F33C8.3
Chromosome Location
Chr.X: 14726626 - 14729503 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078044.7 NP_510445.1 2 - 3 217 70

View Full Product Details