Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-3
Entrez Gene ID
181573
Gene Name
Transcription factor unc-3
Gene Aliases
CELE_Y16B4A.1
Chromosome Location
Chr.X: 14773538 - 14783568 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078052.5 NP_510453.1 7 - 8 814 74
GenBank mRNA AF052060.1 - 7 - 8 844 74

View Full Product Details