Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-3
Entrez Gene ID
181573
Gene Name
Transcription factor unc-3
Gene Aliases
CELE_Y16B4A.1
Chromosome Location
Chr.X: 14773538 - 14783568 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078052.5 NP_510453.1 8 - 9 988 66
GenBank mRNA AF052060.1 - 8 - 9 1018 66

View Full Product Details