Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mbk-1
Entrez Gene ID
181578
Gene Name
Dual specificity tyrosine-phosphorylation-regulated kinase mbk-1
Gene Aliases
CELE_T04C10.1
Chromosome Location
Chr.X: 14819902 - 14824595 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078059.6 NP_510460.2 5 - 6 808 88
GenBank mRNA AY064464.1 - 5 - 6 821 88

View Full Product Details