Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dpf-2
Entrez Gene ID
181579
Gene Name
Dipeptidyl peptidase family member 2
Gene Aliases
CELE_C27C12.7
Chromosome Location
Chr.X: 14831824 - 14838197 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078060.10 NP_510461.1 9 - 10 1692 145

View Full Product Details