Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hex-5
Entrez Gene ID
181586
Gene Name
Glyco_hydro_20 domain-containing protein
Gene Aliases
CELE_Y70D2A.2
Chromosome Location
Chr.X: 14901881 - 14905261 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381161.1 NP_001366664.1 2 - 3 211 136

View Full Product Details