Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pag-3
Entrez Gene ID
181588
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F45B8.4
Chromosome Location
Chr.X: 14950752 - 14954648 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078079.6 NP_510480.1 7 - 8 836 61
GenBank mRNA U63996.1 - 7 - 8 834 61

View Full Product Details