Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs-10
Entrez Gene ID
185764
Gene Name
Regulator of G-protein signaling rgs-10
Gene Aliases
CELE_F45B8.2
Chromosome Location
Chr.X: 14962756 - 14964143 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078081.2 NP_510482.1 3 - 4 763 68

View Full Product Details