Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dsc-1
Entrez Gene ID
181599
Gene Name
Homeobox protein dsc-1
Gene Aliases
CELE_C18B12.3
Chromosome Location
Chr.X: 15031180 - 15034832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078096.4 NP_510497.1 4 - 5 772 90

View Full Product Details