Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-7
Entrez Gene ID
181608
Gene Name
protein-coding
Gene Aliases
CELE_R07D5.1
Chromosome Location
Chr.X: 15120846 - 15147742 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270326.3 NP_001257255.1 2 - 3 277 67
NM_001350358.2 NP_001337320.1 2 - 3 277 67
NM_001375177.1 NP_001361907.1 2 - 3 279 67
GenBank mRNA Z19122.1 - - 298 67

View Full Product Details