Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-7
Entrez Gene ID
181608
Gene Name
protein-coding
Gene Aliases
CELE_R07D5.1
Chromosome Location
Chr.X: 15120846 - 15147742 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270326.3 NP_001257255.1 8 - 9 1216 65
NM_001270327.3 NP_001257256.1 6 - 7 935 65
NM_001350352.4 NP_001337313.1 7 - 7 1205 65
NM_001350353.4 NP_001337312.1 7 - 7 1211 65
NM_001350354.3 NP_001337317.1 7 - 8 1034 65
NM_001350355.3 NP_001337316.1 7 - 8 962 65
NM_001350356.3 NP_001337319.1 10 - 10 1386 65
NM_001350357.3 NP_001337318.1 7 - 8 1001 65
NM_001350358.2 NP_001337320.1 8 - 9 1210 65
NM_001375177.1 NP_001361907.1 8 - 9 1212 65
GenBank mRNA Z19122.1 - - 1237 65

View Full Product Details