Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ocr-3
Entrez Gene ID
188364
Gene Name
ANK_REP_REGION domain-containing protein
Gene Aliases
CELE_T10B10.7
Chromosome Location
Chr.X: 15168722 - 15173636 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078119.7 NP_510520.4 13 - 14 2111 143

View Full Product Details