Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
phat-6
Entrez Gene ID
181614
Gene Name
PHAryngeal gland Toxin-related
Gene Aliases
CELE_T10B10.6
Chromosome Location
Chr.X: 15193086 - 15194383 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078124.6 NP_510525.1 3 - 4 548 72

View Full Product Details